CAS RN 78471-43-9
IUPAC Name:
methyl 4-bromo-2-(bromomethyl)benzoate
Synonyms:
methyl 4-bromo-2-(bromomethyl)benzoate
Molecular Weight (g/mol):
307.97
Molecular Formula:
C9H8Br2O2
InChi Key:
SGFACFBLUAWICV-UHFFFAOYSA-N
SMILES:
COC(=O)C1=C(CBr)C=C(Br)C=C1
1
–
1
of
1
results
1
Methyl 4-bromo-2-(bromomethyl)benzoate, 96%, Thermo Scientific™
CAS: 78471-43-9 Molecular Formula: C9H8Br2O2 Molecular Weight (g/mol): 307.969 MDL Number: MFCD04114319 InChI Key: SGFACFBLUAWICV-UHFFFAOYSA-N Synonym: methyl 4-bromo-2-bromomethyl benzoate,methyl 4-bromo-2-bromomethyl-benzoate,4-bromo-2-bromomethylbenzoic acid methyl ester,4-bromo-2-bromomethyl-benzoic acid methyl ester,benzoic acid, 4-bromo-2-bromomethyl-, methyl ester,4-bromo-2-bromomethyl benzoic acid methyl ester,acmc-209pec,methyl alpha,5-dibromo-o-toluate,methyl 4bromo-2-bromomethylbenzoate,methyl4-bromo-2-bromomethylbenzoate PubChem CID: 22031079 IUPAC Name: methyl 4-bromo-2-(bromomethyl)benzoate SMILES: COC(=O)C1=C(C=C(C=C1)Br)CBr