CAS RN 78239-71-1
CAS RN 78239-71-1
IUPAC Name:
2-bromo-4-(bromomethyl)-1-fluorobenzene
Synonyms:
2-bromo-4-(bromomethyl)-1-fluorobenzene
Molecular Weight (g/mol):
267.92
Molecular Formula:
C7H5Br2F
InChi Key:
ZRWSODQPUJMFRV-UHFFFAOYSA-N
SMILES:
FC1=C(Br)C=C(CBr)C=C1
1
–
1
of
1
results
1
3-Bromo-4-fluorobenzyl bromide, 97%
CAS: 78239-71-1 Molecular Formula: C7H5Br2F Molecular Weight (g/mol): 267.92 MDL Number: MFCD09037367 InChI Key: ZRWSODQPUJMFRV-UHFFFAOYSA-N Synonym: 3-bromo-4-fluorobenzyl bromide,2-bromo-4-bromomethyl-1-fluorobenzene,3-bromo-4-fluorobenzylbromide,benzene, 2-bromo-4-bromomethyl-1-fluoro,2-bromanyl-4-bromomethyl-1-fluoranyl-benzene,pubchem24015,acmc-209pdt,3-bromo-4-fluoro-benzyl bromide,2-bromo-4-bromomethyl-1-fluoro-benzene,benzene,2-bromo-4-bromomethyl-1-fluoro PubChem CID: 12922710 IUPAC Name: 2-bromo-4-(bromomethyl)-1-fluorobenzene SMILES: FC1=C(Br)C=C(CBr)C=C1