accessibility menu, dialog, popup

UserName

CAS RN 78078-50-9

OOMgBr

CAS RN 78078-50-9

IUPAC Name: magnesium(2+) 2-(1,3-dioxan-2-yl)ethanide bromide
Synonyms: magnesium(2+) 2-(1,3-dioxan-2-yl)ethanide bromide
Molecular Weight (g/mol): 219.36
Molecular Formula: C6H11BrMgO2
InChi Key: SWTDOBDFYXFIPN-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].[CH2-]CC1OCCCO1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)