accessibility menu, dialog, popup

UserName

CH3NH3COOH

CAS RN 78062-03-0

IUPAC Name: 4-(dimethylamino)naphthalene-1-carboxylic acid
Synonyms: 4-(dimethylamino)naphthalene-1-carboxylic acid
Molecular Weight (g/mol): 215.25
Molecular Formula: C13H13NO2
InChi Key: LZQKNRRVTOJUHQ-UHFFFAOYSA-N
SMILES: CN(C)C1=C2C=CC=CC2=C(C=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results