accessibility menu, dialog, popup

UserName

CAS RN 78031-18-2

SBrOH2N

CAS RN 78031-18-2

IUPAC Name: 3-bromothiophene-2-carboxamide
Synonyms: 3-bromothiophene-2-carboxamide
Molecular Weight (g/mol): 206.06
Molecular Formula: C5H4BrNOS
InChi Key: KWXUWNXTRUZHNQ-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(Br)C=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results