CAS RN 77987-49-6
CAS RN 77987-49-6
IUPAC Name:
benzyl N-(2-hydroxyethyl)carbamate
Synonyms:
benzyl N-(2-hydroxyethyl)carbamate
Molecular Weight (g/mol):
195.22
Molecular Formula:
C10H13NO3
InChi Key:
SAGINAGERRNGGV-UHFFFAOYSA-N
SMILES:
OCCNC(=O)OCC1=CC=CC=C1
1
–
1
of
1
results
1
N-Z-Ethanolamine, 98%
CAS: 77987-49-6 Molecular Formula: C10H13NO3 Molecular Weight (g/mol): 195.22 MDL Number: MFCD00191060 InChI Key: SAGINAGERRNGGV-UHFFFAOYSA-N Synonym: benzyl n-2-hydroxyethyl carbamate,z-glycinol,benzyl 2-hydroxyethyl carbamate,n-z-ethanolamine,2-cbz-amino-1-ethanol,benzyl 2-hydroxyethylcarbamate,2-z-amino ethanol,n-2-hydroxyethyl phenylmethoxy carboxamide,n-2-hydroxyethyl carbamic acid benzyl ester PubChem CID: 280458 IUPAC Name: benzyl N-(2-hydroxyethyl)carbamate SMILES: OCCNC(=O)OCC1=CC=CC=C1