accessibility menu, dialog, popup

UserName

CAS RN 7791-07-3

H2ONaOClOOO

CAS RN 7791-07-3

IUPAC Name: sodium hydrate perchlorate
Synonyms: sodium hydrate perchlorate
Molecular Weight (g/mol): 140.45
Molecular Formula: ClH2NaO5
InChi Key: IXGNPUSUVRTQGW-UHFFFAOYSA-M
SMILES: O.[Na+].[O-][Cl](=O)(=O)=O
Molecular Weight (g/mol) 
  • (14)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
  • (4)
Grade 
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (1)
  • (1)
  • (2)
  • (4)
  • (14)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (1)
  • (3)
  • (1)