CAS RN 7790-87-6
CAS RN 7790-87-6
IUPAC Name:
cerium(3+) triiodide
Synonyms:
cerium(3+) triiodide
Molecular Weight (g/mol):
520.83
Molecular Formula:
CeI3
InChi Key:
ZEDZJUDTPVFRNB-UHFFFAOYSA-K
SMILES:
[I-].[I-].[I-].[Ce+3]
1
–
1
of
1
results
1
Cerium(III) iodide, ultra dry, 99.9% (REO)
CAS: 7790-87-6 Molecular Formula: CeI3 MDL Number: MFCD00049423 Synonym: Cerium triiodide