CAS RN 779-84-0
CAS RN 779-84-0
IUPAC Name:
(6Z)-6-[(phenylamino)methylidene]cyclohexa-2,4-dien-1-one
Synonyms:
(6Z)-6-[(phenylamino)methylidene]cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol):
197.24
Molecular Formula:
C13H11NO
InChi Key:
GVHXCXKLYZOBKS-KHPPLWFESA-N
SMILES:
O=C1C=CC=C\C1=C\NC1=CC=CC=C1
1
–
2
of
2
results
1
Salicylideneaniline, 98%
CAS: 779-84-0 Molecular Formula: C13H11NO Molecular Weight (g/mol): 197.237 MDL Number: MFCD00020066 InChI Key: GVHXCXKLYZOBKS-UHFFFAOYSA-N Synonym: n-salicylideneaniline,phenol, 2-e-phenylimino methyl,n-salicylalaniline,2-hydroxybenzalaniline,o-hydroxy benzylidene aniline,acmc-20m6ov,acmc-1bijs,n-o-hydroxybenzylidene aniline,2-e-phenyliminomethyl phenol,phenol,2-phenylimino methyl PubChem CID: 6740751 IUPAC Name: 6-(anilinomethylidene)cyclohexa-2,4-dien-1-one SMILES: C1=CC=C(C=C1)NC=C2C=CC=CC2=O