CAS RN 7789-80-2
CAS RN 7789-80-2
IUPAC Name:
calcium diiodate
Synonyms:
calcium iodate
Molecular Weight (g/mol):
389.88
Molecular Formula:
CaI2O6
InChi Key:
UHWJJLGTKIWIJO-UHFFFAOYSA-L
SMILES:
[Ca++].[O-][I](=O)=O.[O-][I](=O)=O
1
–
1
of
1
results
1
Calcium iodate, pure
CAS: 7789-80-2 Molecular Formula: CaI2O6 Molecular Weight (g/mol): 389.89 InChI Key: UHWJJLGTKIWIJO-UHFFFAOYSA-L Synonym: calcium iodate,lautarite,calcium diiodate,unii-l8mn4y57br,l8mn4y57br,iodic acid hio3 , calcium salt 2:1,autarite,iodic acid, calcium salt,calcium iodate ca io3 2,hsdb 986 PubChem CID: 24619 IUPAC Name: calcium;diiodate SMILES: [O-]I(=O)=O.[O-]I(=O)=O.[Ca+2]