accessibility menu, dialog, popup

UserName

CAS RN 7789-40-4

BrTl

CAS RN 7789-40-4

IUPAC Name: λ¹-thallanylium bromide
Synonyms: thallium(I) bromide
Molecular Weight (g/mol): 284.28
Molecular Formula: BrTl
InChi Key: PGAPATLGJSQQBU-UHFFFAOYSA-M
SMILES: [Br-].[Tl+]
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)