CAS RN 7789-28-8
CAS RN 7789-28-8
IUPAC Name:
λ²-iron(2+) difluoride
Synonyms:
iron(II) fluoride
Molecular Weight (g/mol):
93.84
Molecular Formula:
F2Fe
InChi Key:
FZGIHSNZYGFUGM-UHFFFAOYSA-L
SMILES:
[F-].[F-].[Fe++]
1
–
1
of
1
results
1
Iron(II) fluoride, anhydrous, 98%
CAS: 7789-28-8 Molecular Formula: F2Fe Molecular Weight (g/mol): 93.84 MDL Number: MFCD00016093 InChI Key: FZGIHSNZYGFUGM-UHFFFAOYSA-L IUPAC Name: λ²-iron(2+) difluoride SMILES: [F-].[F-].[Fe++]