accessibility menu, dialog, popup

UserName

FFCu

CAS RN 7789-19-7

IUPAC Name: copper(2+) difluoride
Synonyms: copper(II) fluoride
Molecular Weight (g/mol): 101.54
Molecular Formula: CuF2
InChi Key: GWFAVIIMQDUCRA-UHFFFAOYSA-L
SMILES: [F-].[F-].[Cu++]
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)