accessibility menu, dialog, popup

UserName

CAS RN 7783-96-2

AgI

CAS RN 7783-96-2

IUPAC Name: silver(1+) iodide
Synonyms: silver iodide
Molecular Weight (g/mol): 234.77
Molecular Formula: AgI
InChi Key: MSFPLIAKTHOCQP-UHFFFAOYSA-M
SMILES: [Ag+].[I-]
Molecular Weight (g/mol) 
  • (3)
  • (7)
Physical Form 
  • (2)
  • (3)
  • (3)
  • (2)
Quantity 
  • (4)
  • (4)
  • (2)
Percent Purity 
  • (5)
  • (3)
Grade 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (5)
  • (3)
  • (7)
  • (2)
  • (3)
  • (3)
  • (2)
  • (4)
  • (4)
  • (2)
  • (5)
  • (3)
  • (3)
  • (3)