accessibility menu, dialog, popup

UserName

CAS RN 7783-18-8

IUPAC Name: dihydroxy-1λ⁶-disulfen-1-one diamine
Synonyms: ammonium thiosulfate
Molecular Weight (g/mol): 148.2
Molecular Formula: H8N2O3S2
InChi Key: XYXNTHIYBIDHGM-UHFFFAOYSA-N
SMILES: N.N.OS(O)(=O)=S
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Physical Form 
  • (3)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)