accessibility menu, dialog, popup

UserName

CAS RN 77771-02-9

FOBr

CAS RN 77771-02-9

IUPAC Name: 3-bromo-4-fluorobenzaldehyde
Synonyms: 3-bromo-4-fluorobenzaldehyde
Molecular Weight (g/mol): 203.01
Molecular Formula: C7H4BrFO
InChi Key: FAHZIKXYYRGSHF-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=O)C=C1Br
Boiling Point 
  • (4)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)