CAS RN 7758-11-4
CAS RN 7758-11-4
IUPAC Name:
dipotassium hydrogen phosphate
Synonyms:
dipotassium phosphate potassium phosphate, dibasic
Molecular Weight (g/mol):
174.17
Molecular Formula:
HK2O4P
InChi Key:
ZPWVASYFFYYZEW-UHFFFAOYSA-L
SMILES:
[K+].[K+].OP([O-])([O-])=O
1
–
1
of
1
results
1
Potassium Phosphate, Dibasic, 1.0M solution in water
CAS: 7758-11-4 | HK2O4P | 174.18 g/mol