accessibility menu, dialog, popup

UserName

CAS RN 775351-40-1

BOOH3CCH3H3CH3CNNHNN

CAS RN 775351-40-1

IUPAC Name: 5-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-2H-1,2,3,4-tetrazole
Synonyms: 5-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-2H-1,2,3,4-tetrazole
Molecular Weight (g/mol): 272.12
Molecular Formula: C13H17BN4O2
InChi Key: ONAVJWUKLJDJAI-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C1=NNN=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results