CAS RN 77159-76-3
IUPAC Name:
5-bromo-2-isocyanato-1,3-dimethylbenzene
Synonyms:
5-bromo-2-isocyanato-1,3-dimethylbenzene
Molecular Weight (g/mol):
226.07
Molecular Formula:
C9H8BrNO
InChi Key:
XGBDVSJAHTYSBK-UHFFFAOYSA-N
SMILES:
CC1=CC(Br)=CC(C)=C1N=C=O
1
–
1
of
1
results
1
4-Bromo-2,6-dimethylphenyl isocyanate, 98%, Thermo Scientific™
CAS: 77159-76-3 Molecular Formula: C9H8BrNO Molecular Weight (g/mol): 226.07 MDL Number: MFCD00037040 InChI Key: XGBDVSJAHTYSBK-UHFFFAOYSA-N Synonym: 4-bromo-2,6-dimethylphenyl isocyanate,4-bromo-2,6-dimethylphenylisocyanate,4-bromo-2,6-dimethylbenzenisocyanate,benzene,5-bromo-2-isocyanato-1,3-dimethyl,5-bromanyl-2-isocyanato-1,3-dimethyl-benzene,benzene, 5-bromo-2-isocyanato-1,3-dimethyl-9ci PubChem CID: 4430747 IUPAC Name: 5-bromo-2-isocyanato-1,3-dimethylbenzene SMILES: CC1=CC(Br)=CC(C)=C1N=C=O