CAS RN 77145-61-0
CAS RN 77145-61-0
IUPAC Name:
hydrogen 1-(6-chloropyridin-2-yl)piperidin-4-amine chloride
Synonyms:
hydrogen 1-(6-chloropyridin-2-yl)piperidin-4-amine chloride
Molecular Weight (g/mol):
248.15
Molecular Formula:
C10H15Cl2N3
InChi Key:
FUMINTAAUJUVMP-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].NC1CCN(CC1)C1=CC=CC(Cl)=N1
1
–
1
of
1
results
1
Tocris Bioscience™ SR 57227 hydrochloride
CAS: 77145-61-0 Molecular Formula: C10H15Cl2N3