accessibility menu, dialog, popup

UserName

FFFSHFF

CAS RN 771-62-0

IUPAC Name: pentafluorobenzene-1-thiol
Synonyms: pentafluorothiophenol
Molecular Weight (g/mol): 200.13
Molecular Formula: C6HF5S
InChi Key: UVAMFBJPMUMURT-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(S)C(F)=C1F
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)
  • (3)