accessibility menu, dialog, popup

UserName

CAS RN 77047-87-1

H3CCHCH3ZnBr

CAS RN 77047-87-1

IUPAC Name: bromozincylium; propan-2-ide
Synonyms: bromozincylium; propan-2-ide
Molecular Weight (g/mol): 188.37
Molecular Formula: C3H7BrZn
InChi Key: NTUATXZETUXBSR-UHFFFAOYSA-M
SMILES: [Zn+]Br.C[CH-]C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results