accessibility menu, dialog, popup

UserName

CAS RN 770-71-8

IUPAC Name: (adamantan-1-yl)methanol
Synonyms: adamantan-1-ylmethanol
Molecular Weight (g/mol): 166.26
Molecular Formula: C11H18O
InChi Key: MDVGOOIANLZFCP-UHFFFAOYSA-N
SMILES: OCC12CC3CC(CC(C3)C1)C2
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)