accessibility menu, dialog, popup

UserName

CAS RN 77-95-2

IUPAC Name: 1,3,4,5-tetrahydroxycyclohexane-1-carboxylic acid
Synonyms: quinic acid
Molecular Weight (g/mol): 192.17
Molecular Formula: C7H12O6
InChi Key: AAWZDTNXLSGCEK-UHFFFAOYNA-N
SMILES: OC1CC(O)(CC(O)C1O)C(O)=O
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)