CAS RN 77-38-3
CAS RN 77-38-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Chlorphenoxamine, TRC
CAS: 77-38-3 Molecular Formula: C18 H22 Cl N O Molecular Weight (g/mol): 303.83 IUPAC Name: 2-[1-(4-chlorophenyl)-1-phenyl-ethoxy]-N,N-dimethyl-ethanamine SMILES: CN(C)CCOC(C)(c1ccccc1)c2ccc(Cl)cc2