accessibility menu, dialog, popup

UserName

CAS RN 7697-23-6

CH3OHOF

CAS RN 7697-23-6

IUPAC Name: 2-fluoro-4-methylbenzoic acid
Synonyms: 2-fluoro-4-methylbenzoic acid
Molecular Weight (g/mol): 154.14
Molecular Formula: C8H7FO2
InChi Key: ALFWHEYHCZRVLO-UHFFFAOYSA-N
SMILES: CC1=CC=C(C(O)=O)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results