accessibility menu, dialog, popup

UserName

CAS RN 769-92-6

CH3H3CCH3NH2

CAS RN 769-92-6

IUPAC Name: 4-tert-butylaniline
Synonyms: 4-tert-butylaniline
Molecular Weight (g/mol): 149.24
Molecular Formula: C10H15N
InChi Key: WRDWWAVNELMWAM-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(N)C=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (4)
  • (2)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)