accessibility menu, dialog, popup

UserName

CH3NOHNCH3

CAS RN 769-28-8

IUPAC Name: 4,6-dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile
Synonyms: 4,6-dimethyl-2-oxo-1H-pyridine-3-carbonitrile
Molecular Weight (g/mol): 148.17
Molecular Formula: C8H8N2O
InChi Key: OCYMJCILWYHKAU-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C#N)C(=O)N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results