accessibility menu, dialog, popup

UserName

CAS RN 769-10-8

CH3FNOO

CAS RN 769-10-8

IUPAC Name: 1-fluoro-2-methyl-3-nitrobenzene
Synonyms: 1-fluoro-2-methyl-3-nitrobenzene
Molecular Weight (g/mol): 155.13
Molecular Formula: C7H6FNO2
InChi Key: GXPIVRKDWZKIKZ-UHFFFAOYSA-N
SMILES: CC1=C(F)C=CC=C1[N+]([O-])=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results