accessibility menu, dialog, popup

UserName

CAS RN 766-80-3

ClBr

CAS RN 766-80-3

IUPAC Name: 1-(bromomethyl)-3-chlorobenzene
Synonyms: 1-(bromomethyl)-3-chlorobenzene
Molecular Weight (g/mol): 205.48
Molecular Formula: C7H6BrCl
InChi Key: LZIYAIRGDHSVED-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(CBr)=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)