accessibility menu, dialog, popup

UserName

CAS RN 76575-71-8

CH3SOOH3CNH2

CAS RN 76575-71-8

IUPAC Name: methyl 3-amino-5-methylthiophene-2-carboxylate
Synonyms: methyl 3-amino-5-methylthiophene-2-carboxylate
Molecular Weight (g/mol): 171.21
Molecular Formula: C7H9NO2S
InChi Key: FVKMOPIFLCMZMI-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(N)C=C(C)S1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results