accessibility menu, dialog, popup

UserName

CAS RN 764-42-1

NN(E)

CAS RN 764-42-1

IUPAC Name: (2E)-but-2-enedinitrile
Synonyms: 2-butenedinitrile, (E)-
Molecular Weight (g/mol): 78.07
Molecular Formula: C4H2N2
InChi Key: KYPOHTVBFVELTG-OWOJBTEDSA-N
SMILES: N#C\C=C\C#N
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)