accessibility menu, dialog, popup

UserName

CAS RN 7631-95-0

NaNaOMoOOO

CAS RN 7631-95-0

IUPAC Name: disodium dioxomolybdenumbis(olate)
Synonyms: sodium molybdate
Molecular Weight (g/mol): 205.93
Molecular Formula: MoNa2O4
InChi Key: TVXXNOYZHKPKGW-UHFFFAOYSA-N
SMILES: [Na+].[Na+].[O-][Mo]([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)