accessibility menu, dialog, popup

UserName

CAS RN 76041-73-1

IUPAC Name: 3-bromo-5-(trifluoromethyl)-1,2-dihydropyridin-2-one
Synonyms: 3-bromo-5-(trifluoromethyl)-1H-pyridin-2-one
Molecular Weight (g/mol): 242
Molecular Formula: C6H3BrF3NO
InChi Key: UWHKLLYQUQPOQK-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CNC(=O)C(Br)=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results