accessibility menu, dialog, popup

UserName

NaNaNaOPOOO

CAS RN 7601-54-9

IUPAC Name: trisodium phosphate
Synonyms: sodium phosphate tertiary sodium phosphate tertiaeres natriumphosphat(V) tribasic sodium orthophosphate Na3PO4 phosphoric acid trisodium salt trinatriumphosphat tribasic sodium phosphate sodium phosphate, tribasic sodium orthophosphate
Molecular Weight (g/mol): 163.94
Molecular Formula: Na3O4P
InChi Key: RYFMWSXOAZQYPI-UHFFFAOYSA-K
SMILES: [Na+].[Na+].[Na+].[O-]P([O-])([O-])=O
Grade 
  • (3)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Physical Form 
  • (1)
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)