accessibility menu, dialog, popup

UserName

NClBrFFF

CAS RN 75806-84-7

IUPAC Name: 2-bromo-3-chloro-5-(trifluoromethyl)pyridine
Synonyms: 2-bromo-3-chloro-5-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 260.44
Molecular Formula: C6H2BrClF3N
InChi Key: SMTKGMYGLYWNDL-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Cl)=C(Br)N=C1
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results