accessibility menu, dialog, popup

UserName

H3CCH3H3COON

CAS RN 75400-67-8

IUPAC Name: tert-butyl 1H-indole-1-carboxylate
Synonyms: tert-butyl indole-1-carboxylate
Molecular Weight (g/mol): 217.27
Molecular Formula: C13H15NO2
InChi Key: OWPIFQXNMLDXKW-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1C=CC2=CC=CC=C12
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)