accessibility menu, dialog, popup

UserName

CAS RN 754-91-6

NH2SOOFFFFFFFFFFFFFFFFF

CAS RN 754-91-6

IUPAC Name: heptadecafluorooctane-1-sulfonamide
Synonyms: perfluorooctanesulfonamide
Molecular Weight (g/mol): 499.14
Molecular Formula: C8H2F17NO2S
InChi Key: RRRXPPIDPYTNJG-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results