accessibility menu, dialog, popup

UserName

CAS RN 75-94-5

ClSiClClH2C

CAS RN 75-94-5

IUPAC Name: trichloro(ethenyl)silane
Synonyms: vinyltrichlorosilane
Molecular Weight (g/mol): 161.48
Molecular Formula: C2H3Cl3Si
InChi Key: GQIUQDDJKHLHTB-UHFFFAOYSA-N
SMILES: Cl[Si](Cl)(Cl)C=C
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)