CAS RN 74863-84-6
CAS RN 74863-84-6
IUPAC Name:
(2R,4R)-1-{5-[(diaminomethylidene)amino]-2-(3-methyl-1,2,3,4-tetrahydroquinoline-8-sulfonamido)pentanoyl}-4-methylpiperidine-2-carboxylic acid
Synonyms:
argatroban
Molecular Weight (g/mol):
508.64
Molecular Formula:
C23H36N6O5S
InChi Key:
KXNPVXPOPUZYGB-VSVYTNTFSA-N
SMILES:
C[C@@H]1CCN([C@H](C1)C(O)=O)C(=O)C(CCCN=C(N)N)NS(=O)(=O)C1=CC=CC2=C1NCC(C)C2
1
–
5
of
5
results
1
Thermo Scientific Chemicals Argatroban
CAS: 74863-84-6 Molecular Formula: C23H36N6O5S
2
Argatroban, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Tocris Bioscience™ Argatroban
Potent thrombin inhibitor
4
Argatroban, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
5
Argatroban, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.