CAS RN 740846-54-2
CAS RN 740846-54-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Formyl-N-propyl-N-[2-(2,4,6-trichlorophenoxy)ethyl]-hydrazinecarboxamide, TRC
CAS: 740846-54-2 Molecular Formula: C13 H16 Cl3 N3 O3 Molecular Weight (g/mol): 368.64 Synonym: 2-Formyl-N-propyl-N-[2-(2,4,6-trichlorophenoxy)ethyl]-hydrazinecarboxamide,2-Formyl-N-propyl-N-[2-(2,4,6-trichlorophenoxy)ethyl]-hydrazinecarboxamide IUPAC Name: N-[[propyl-[2-(2,4,6-trichlorophenoxy)ethyl]carbamoyl]amino]formamide SMILES: CCCN(CCOc1c(Cl)cc(Cl)cc1Cl)C(=O)NNC=O