accessibility menu, dialog, popup

UserName

H3CNHCH3BH3

CAS RN 74-94-2

IUPAC Name: dimethylamine boron
Synonyms: dimethylamine boron
Molecular Weight (g/mol): 55.9
Molecular Formula: C2H7BN
InChi Key: RJTANRZEWTUVMA-UHFFFAOYSA-N
SMILES: [B].CNC
Percent Purity 
  • (2)
  • (4)
Physical Form 
  • (2)
  • (4)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)