accessibility menu, dialog, popup

UserName

CAS RN 74-39-5

NHNOOHNOO(E/Z)

CAS RN 74-39-5

IUPAC Name: 3-hydroxy-4-[2-(4-nitrophenyl)hydrazin-1-ylidene]cyclohexa-2,5-dien-1-one
Synonyms: azo violet
Molecular Weight (g/mol): 259.22
Molecular Formula: C12H9N3O4
InChi Key: BIFFKKLAAIGZBV-UHFFFAOYSA-N
SMILES: OC1=CC(=O)C=CC1=NNC1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)