accessibility menu, dialog, popup

UserName

H3CNNNOOH

CAS RN 739364-95-5

IUPAC Name: 2-methylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid
Synonyms: 2-methylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid
Molecular Weight (g/mol): 177.16
Molecular Formula: C8H7N3O2
InChi Key: YHVGUXFUOSJCFJ-UHFFFAOYSA-N
SMILES: CC1=NN2C=C(C=NC2=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results