accessibility menu, dialog, popup

UserName

CAS RN 7312-24-5

BrSOOH

CAS RN 7312-24-5

IUPAC Name: 5-bromo-1-benzothiophene-3-carboxylic acid
Synonyms: 5-bromo-1-benzothiophene-3-carboxylic acid
Molecular Weight (g/mol): 257.1
Molecular Formula: C9H5BrO2S
InChi Key: DWQBOPASRUUSKR-UHFFFAOYSA-N
SMILES: OC(=O)C1=CSC2=CC=C(Br)C=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results