accessibility menu, dialog, popup

UserName

CAS RN 72804-96-7

POOH2N

CAS RN 72804-96-7

IUPAC Name: amino diphenylphosphinate
Synonyms: amino diphenylphosphinate
Molecular Weight (g/mol): 233.21
Molecular Formula: C12H12NO2P
InChi Key: SJECIYLGISUNRO-UHFFFAOYSA-N
SMILES: NOP(=O)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results