accessibility menu, dialog, popup

UserName

CAS RN 72754-05-3

IUPAC Name: 6-bromo-1,2-dihydro-1,8-naphthyridin-2-one
Synonyms: 6-bromo-1H-1,8-naphthyridin-2-one
Molecular Weight (g/mol): 225.05
Molecular Formula: C8H5BrN2O
InChi Key: BCEWGGGNOQNYKK-UHFFFAOYSA-N
SMILES: BrC1=CN=C2NC(=O)C=CC2=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results