accessibility menu, dialog, popup

UserName

CH3O(S)O(S)CH3OH3CHOONHNCH3H3CNONHO(S)H3C(S)HO(R)H3C(R)HO(R)H3C(S)OCH3O(R)H3CCH3(?)(Z)(Z)

CAS RN 72559-06-9

IUPAC Name: (7S,9E,11S,12R,13S,14R,15R,16R,17S,18S,19E,21Z)-2,15,17-trihydroxy-11-methoxy-3,7,12,14,16,18,22-heptamethyl-1'-(2-methylpropyl)-6,23,32-trioxo-8,33-dioxa-24,27,29-triazaspiro[pentacyclo[23.6.1.1⁴,⁷.0⁵,³¹.0²⁶,³⁰]tritriacontane-28,4'-piperidin]-1,3,5(31),9,19,21,25,29-octaen-13-yl acetate
Synonyms: rifabutin 利福布汀
Molecular Weight (g/mol): 847.02
Molecular Formula: C46H62N4O11
InChi Key: ATEBXHFBFRCZMA-VXTBVIBXSA-N
SMILES: CO[C@H]1\C=C\O[C@@]2(C)OC3=C(C)C(O)=C4C(=O)C(NC(=O)\C(C)=C/C=C/[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(C)=O)[C@@H]1C)=C1NC5(CCN(CC(C)C)CC5)N=C1C4=C3C2=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results