accessibility menu, dialog, popup

UserName

NHOF

CAS RN 725-38-2

IUPAC Name: N-benzyl-4-fluorobenzamide
Synonyms: N-benzyl-4-fluorobenzamide
Molecular Weight (g/mol): 229.25
Molecular Formula: C14H12FNO
InChi Key: XCRBOSJQBAFSMZ-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C(=O)NCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results