CAS RN 7238-62-2
CAS RN 7238-62-2
IUPAC Name:
1-chloro-3-[4-nitro-5-(piperidin-1-yl)-1H-imidazol-1-yl]propan-2-ol
Synonyms:
1-chloro-3-[4-nitro-5-(piperidin-1-yl)imidazol-1-yl]propan-2-ol
Molecular Weight (g/mol):
288.73
Molecular Formula:
C11H17ClN4O3
InChi Key:
PKWGLVDKUPRXEN-UHFFFAOYNA-N
SMILES:
OC(CCl)CN1C=NC(=C1N1CCCCC1)[N+]([O-])=O
1
–
1
of
1
results
1
Ethyl 2-chloro-4-methyl-1,3-thiazole-5-carboxylate, 97%, Thermo Scientific™
CAS: 7238-62-2 Molecular Formula: C7H8ClNO2S Molecular Weight (g/mol): 205.656 InChI Key: VUARUZUFHDNJSY-UHFFFAOYSA-N Synonym: ethyl 2-chloro-4-methylthiazole-5-carboxylate,ethyl 2-chloro-4-methyl-5-thiazolecarboxylate,5-thiazolecarboxylic acid, 2-chloro-4-methyl-, ethyl ester,2-chloro-4-methyl-5-thiazolecarboxylic acid ethyl ester,5-thiazolecarboxylicacid, 2-chloro-4-methyl-, ethyl ester,2-chloro-4-methyl-thiazole-5-carboxylic acid ethyl ester PubChem CID: 594903 IUPAC Name: ethyl 2-chloro-4-methyl-1,3-thiazole-5-carboxylate SMILES: CCOC(=O)C1=C(N=C(S1)Cl)C